MAN412114504 Manchester Organics

N-[3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluoro-phenyl]propane-1-sulfonamide

  • MDL number MFCD14635203
  • Molecular Weight : 413.826

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E35931
Availability details for
Quantity
  • Availability
  • Add to Cart