MAN412114532 Manchester Organics

N-(2-Aminophenyl)-4-[[[4-(3-pyridyl)pyrimidin-2-yl]amino]methyl]benzamide

  • MDL number MFCD10565970
  • Cas Number : 726169-73-9
  • Molecular Weight : 396.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/K35959
Availability details for
Quantity
  • Availability
  • Add to Cart