MAN412114555 Manchester Organics

5-Methyl-2-nitro-pyridine

  • MDL number MFCD00234182
  • Cas Number : 1074-38-0
  • Molecular Weight : 138.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M35984
Availability details for
Quantity
  • Availability
  • Add to Cart