MAN412114934 Manchester Organics

Methyl 2-aminopyrimidine-5-carboxylate

  • MDL number MFCD08275693
  • Molecular Weight : 153.13868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B36411

Safety Information

MSDS https://www.manchesterorganics.com/B36411
Availability details for
Quantity
  • Availability
  • Add to Cart