MAN412114962 Manchester Organics

2,4-Diaminopyrimidine-5-carbonitrile

  • MDL number MFCD00085600
  • Cas Number : 16462-27-4
  • Molecular Weight : 135.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/G36439
Availability details for
Quantity
  • Availability
  • Add to Cart