MAN412114968 Manchester Organics

Methyl 5-chloro-4-nitro-thiophene-2-carboxylate

  • MDL number MFCD08275718
  • Molecular Weight : 221.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin GB
Product Link https://www.manchesterorganics.com/N36445
Availability details for
Quantity
  • Availability
  • Add to Cart