MAN412115123 Manchester Organics

tert-Butyl 4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylate

  • MDL number MFCD08061257
  • Cas Number : 657428-42-7
  • Molecular Weight : 209.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T36702
Availability details for
Quantity
  • Availability
  • Add to Cart