MAN412115127 Manchester Organics

4-Aminopyridin-2-ol

  • MDL number MFCD02682913
  • Molecular Weight : 110.114

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X36706
Availability details for
Quantity
  • Availability
  • Add to Cart