MAN412115139 Manchester Organics

4-Chloro-6-methoxypyrimidine

  • MDL number MFCD06200211
  • Cas Number : 26452-81-3
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-pyrimidinyl methyl ether
Inchi Code 1S/C5H5ClN2O/c1-9-5-2-4(6)7-3-8-5/h2-3H,1H3
Inchi Key KLJGSQVYUGQOAW-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/K36718
Availability details for
Quantity
  • Availability
  • Add to Cart