MAN412115216 Manchester Organics

2-Amino-5-bromo-pyridin-3-ol

  • MDL number MFCD09744143
  • Cas Number : 39903-01-0
  • Molecular Weight : 189.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-5-bromo-3-pyridinol
Inchi Code 1S/C5H5BrN2O/c6-3-1-4(9)5(7)8-2-3/h1-2,9H,(H2,7,8)
Inchi Key YQADLKDQAXAIKW-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/U36795
Availability details for
Quantity
  • Availability
  • Add to Cart