MAN412115262 Manchester Organics

5-Methoxypyrazin-2-amine

  • MDL number MFCD09909647
  • Cas Number : 54013-07-9
  • Molecular Weight : 125.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U36841
Availability details for
Quantity
  • Availability
  • Add to Cart