MAN412115272 Manchester Organics

5-Bromopyrimidin-4-ol

  • MDL number MFCD00223740
  • Cas Number : 19808-30-1
  • Molecular Weight : 174.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/E36851
Availability details for
Quantity
  • Availability
  • Add to Cart