MAN412115428 Manchester Organics

4H-Isoquinoline-1,3-dione

  • MDL number MFCD00234966
  • Molecular Weight : 161.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z37007
Availability details for
Quantity
  • Availability
  • Add to Cart