MAN412115555 Manchester Organics

4-Hydroxy-2-methyl-benzaldehyde

  • MDL number MFCD04037443
  • Cas Number : 41438-18-0
  • Molecular Weight : 136.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M37134
Availability details for
Quantity
  • Availability
  • Add to Cart