MAN412115574 Manchester Organics

3,5-Dichloropyrazin-2-amine

  • MDL number MFCD11040179
  • Cas Number : 873-42-7
  • Molecular Weight : 163.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/H37153
Availability details for
Quantity
  • Availability
  • Add to Cart