MAN412115631 Manchester Organics

6-Bromopyrimidin-4-amine

  • MDL number MFCD00233963
  • Cas Number : 1159818-57-1
  • Molecular Weight : 174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/V37210
Availability details for
Quantity
  • Availability
  • Add to Cart