MAN412115644 Manchester Organics

4,6-Dichloro-2-methyl-5-nitropyrimidine

  • MDL number MFCD00060527
  • Molecular Weight : 208.002

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J37223
Availability details for
Quantity
  • Availability
  • Add to Cart