MAN412115680 Manchester Organics

(3S)-4-tert-butoxycarbonylmorpholine-3-carboxylic acid

  • MDL number MFCD04114904
  • Cas Number : 783350-37-8
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S)-4-(tert-butoxycarbonyl)-3-morpholinecarboxylic acid
Inchi Code 1S/C10H17NO5/c1-10(2,3)16-9(14)11-4-5-15-6-7(11)8(12)13/h7H,4-6H2,1-3H3,(H,12,13)/t7-/m0/s1
Inchi Key KVXXEKIGMOEPSA-ZETCQYMHSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y37259
Availability details for
Quantity
  • Availability
  • Add to Cart