MAN412115721 Manchester Organics

2-Bromo-6-chloro-benzonitrile

  • MDL number MFCD06335184
  • Cas Number : 6575-08-2
  • Molecular Weight : 216.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-6-chlorobenzonitrile
Inchi Code 1S/C7H3BrClN/c8-6-2-1-3-7(9)5(6)4-10/h1-3H
Inchi Key NVPXACXEVCJWCF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/T37300
Availability details for
Quantity
  • Availability
  • Add to Cart