MAN412115803 Manchester Organics

(10-Phenyl-9-anthryl)boronic acid

  • MDL number MFCD11111989
  • Molecular Weight : 298.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.98
Country of Origin GB
Product Link https://www.manchesterorganics.com/K37707
Availability details for
Quantity
  • Availability
  • Add to Cart