MAN412116165 Manchester Organics

(2-Pyrrolidin-1-ylphenyl)methanamine

  • MDL number MFCD04971089
  • Cas Number : 72752-53-5
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-pyrrolidinyl)benzylamine
Inchi Code 1S/C11H16N2/c12-9-10-5-1-2-6-11(10)13-7-3-4-8-13/h1-2,5-6H,3-4,7-9,12H2
Inchi Key PLXNYYYHSZRVJH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/E38070
Availability details for
Quantity
  • Availability
  • Add to Cart