MAN412116174 Manchester Organics

(2R,3R,4R,5R)-2-(4-Aminopyrrolo[2,3-d]pyrimidin-7-yl)-5-(hydroxymethyl)-3-methyl-tetrahydrofuran-3,4-diol

  • MDL number MFCD14584964
  • Cas Number : 443642-29-3
  • Molecular Weight : 280.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P38079
Availability details for
Quantity
  • Availability
  • Add to Cart