MAN412116189 Manchester Organics

(2S)-2-Amino-N-(4-methyl-2-oxo-chromen-7-yl)propanamide

  • MDL number MFCD00077086
  • Molecular Weight : 246.262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F38094
Availability details for
Quantity
  • Availability
  • Add to Cart