MAN412116894 Manchester Organics

2-[(2R)-1-tert-butoxycarbonyl-2-piperidyl]acetic acid

  • MDL number MFCD06656438
  • Cas Number : 351410-32-7
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y38800
Availability details for
Quantity
  • Availability
  • Add to Cart