MAN412117138 Manchester Organics

(3R)-3-Methylpyrrolidine hydrochloride

  • MDL number MFCD06796631
  • Cas Number : 235093-98-8
  • Molecular Weight : 121.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/N39044
Availability details for
Quantity
  • Availability
  • Add to Cart