MAN412117249 Manchester Organics

tert-Butyl (2R)-2-(Bromomethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD12912139
  • Cas Number : 1039826-29-3
  • Molecular Weight : 264.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/J39155
Availability details for
Quantity
  • Availability
  • Add to Cart