MAN412117402 Manchester Organics

(2S)-2-Amino-3-hydroxy-3-methyl-butanoic acid

  • MDL number MFCD00800535
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M39526
Availability details for
Quantity
  • Availability
  • Add to Cart