MAN412117406 Manchester Organics

(2S)-2-Aminohept-6-enoic acid

  • MDL number MFCD06200653
  • Cas Number : 166734-64-1
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2S)-2-amino-6-heptenoic acid
Inchi Code 1S/C7H13NO2/c1-2-3-4-5-6(8)7(9)10/h2,6H,1,3-5,8H2,(H,9,10)/t6-/m0/s1
Inchi Key OOOVDVSHGOKTNT-LURJTMIESA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/S39530
Availability details for
Quantity
  • Availability
  • Add to Cart