MAN412117463 Manchester Organics

tert-Butyl (2S)-2-(methylaminomethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD07786221
  • Cas Number : 191231-58-0
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/D39587
Availability details for
Quantity
  • Availability
  • Add to Cart