MAN412117589 Manchester Organics

(4S)-4-Isobutyloxazolidine-2,5-dione

  • MDL number MFCD03411292
  • Cas Number : 3190-70-3
  • Molecular Weight : 157.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R39713
Availability details for
Quantity
  • Availability
  • Add to Cart