MAN412117790 Manchester Organics

3-Fluoropyridine-4-carbonitrile

  • MDL number MFCD04112524
  • Cas Number : 113770-88-0
  • Molecular Weight : 122.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoroisonicotinonitrile
Inchi Code 1S/C6H3FN2/c7-6-4-9-2-1-5(6)3-8/h1-2,4H
Inchi Key RFQHDTRSEWCXJP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C39908
Availability details for
Quantity
  • Availability
  • Add to Cart