MAN412118082 Manchester Organics

Prop-1-ene-1,3-sultone

  • MDL number MFCD12405143
  • Molecular Weight : 120.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z40204
Availability details for
Quantity
  • Availability
  • Add to Cart