MAN412118212 Manchester Organics

4-Cyanopiperidine hydrochloride

  • MDL number MFCD09037930
  • Molecular Weight : 146.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H15237
Availability details for
Quantity
  • Availability
  • Add to Cart