MAN412118215 Manchester Organics

2-Chloro-1,3-benzothiazol-6-amine

  • MDL number MFCD08276901
  • Cas Number : 2406-90-8
  • Molecular Weight : 184.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-1,3-benzothiazol-6-amine
Inchi Code 1S/C7H5ClN2S/c8-7-10-5-2-1-4(9)3-6(5)11-7/h1-3H,9H2
Inchi Key YPTWPDOGEAHMOR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/V11084
Availability details for
Quantity
  • Availability
  • Add to Cart