MAN412118219 Manchester Organics

3-Methylthiophene-2-carbonitrile

  • MDL number MFCD00052086
  • Molecular Weight : 123.1756

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T39899

Safety Information

MSDS https://www.manchesterorganics.com/T39899
Availability details for
Quantity
  • Availability
  • Add to Cart