MAN412118220 Manchester Organics

4-Hydroxycyclohexanecarboxylic acid

  • MDL number MFCD00077974
  • Molecular Weight : 144.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T40336
Availability details for
Quantity
  • Availability
  • Add to Cart