MAN412118256 Manchester Organics

3-Methylthiophene-2-carboxamide

  • MDL number MFCD01208055
  • Cas Number : 76655-99-7
  • Molecular Weight : 141.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W11085
Availability details for
Quantity
  • Availability
  • Add to Cart