MAN412118261 Manchester Organics

6-Aminopyridazine-3-carboxylic acid

  • MDL number MFCD06658328
  • Cas Number : 59772-58-6
  • Molecular Weight : 139.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/H40373
Availability details for
Quantity
  • Availability
  • Add to Cart