MAN412118353 Manchester Organics

Ethyl 2-chlorooxazole-5-carboxylate

  • MDL number MFCD09475866
  • Cas Number : 862599-47-1
  • Molecular Weight : 175.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-chloro-1,3-oxazole-5-carboxylate
Inchi Code 1S/C6H6ClNO3/c1-2-10-5(9)4-3-8-6(7)11-4/h3H,2H2,1H3
Inchi Key ZFLDTEIDAGWEEI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y40456
Availability details for
Quantity
  • Availability
  • Add to Cart