MAN412118393 Manchester Organics

3-Bromo-6-methyl-pyridin-2-ol

  • MDL number MFCD03095217
  • Cas Number : 374633-33-7
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/N40493
Availability details for
Quantity
  • Availability
  • Add to Cart