MAN412118434 Manchester Organics

3-(Difluoromethoxy)-2-fluoro-pyridine

  • MDL number MFCD13185836
  • Molecular Weight : 163.09726

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A14687

Safety Information

MSDS https://www.manchesterorganics.com/A14687
Availability details for
Quantity
  • Availability
  • Add to Cart