MAN412118445 Manchester Organics

2-Chloropyrimidin-5-ol

  • MDL number MFCD09743796
  • Cas Number : 4983-28-2
  • Molecular Weight : 130.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/U40544
Availability details for
Quantity
  • Availability
  • Add to Cart