MAN412118493 Manchester Organics

2-Methoxy-2-methyl-propan-1-amine

  • MDL number MFCD01821229
  • Molecular Weight : 103.16281

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W40592

Safety Information

MSDS https://www.manchesterorganics.com/W40592
Availability details for
Quantity
  • Availability
  • Add to Cart