MAN412118555 Manchester Organics

Dimethyl hydrogen phosphate

  • MDL number MFCD00014887
  • Molecular Weight : 126.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/P40655
Availability details for
Quantity
  • Availability
  • Add to Cart