MAN412118592 Manchester Organics

Nonyl(triphenyl)phosphonium bromide

  • MDL number MFCD00051875
  • Molecular Weight : 469.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E40692
Availability details for
Quantity
  • Availability
  • Add to Cart