MAN412118666 Manchester Organics

(3-Ethoxy-3-oxo-propyl)-triphenyl-phosphonium bromide

  • MDL number MFCD03701355
  • Molecular Weight : 443.313

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K40766
Availability details for
Quantity
  • Availability
  • Add to Cart