MAN412118712 Manchester Organics

1-Dimethoxyphosphoryl-3-phenyl-propan-2-one

  • MDL number MFCD09951913
  • Molecular Weight : 242.208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.manchesterorganics.com/product.php/K40812
Availability details for
Quantity
  • Availability
  • Add to Cart