MAN412118973 Manchester Organics

tert-Butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate

  • MDL number MFCD04221008
  • Cas Number : 127589-93-9
  • Molecular Weight : 229.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/D41059
Availability details for
Quantity
  • Availability
  • Add to Cart