MAN412119046 Manchester Organics

Isobutyl methanesulfonate

  • MDL number MFCD07363982
  • Molecular Weight : 152.21197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H41132

Safety Information

MSDS https://www.manchesterorganics.com/H41132
Availability details for
Quantity
  • Availability
  • Add to Cart