MAN412119087 Manchester Organics

2-Hydroxy-4-nitrobenzaldehyde

  • MDL number MFCD00049373
  • Cas Number : 2460-58-4
  • Molecular Weight : 167.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/C41173
Availability details for
Quantity
  • Availability
  • Add to Cart